Product Details

SNP ID
rs148864550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238239146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGGAGCTCGGCAGCGACGGTA[C/G]CGTCGATGGCCTGGCACGTGGACAC
Phenotype
MIM: 610331 MIM: 603426
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HES6 PubMed Links

Gene Details

Gene
HES6
Gene Name
hes family bHLH transcription factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142853.2 569 Missense Mutation GCT,GGT A117G NP_001136325.1
NM_001282434.1 569 Missense Mutation CTA,GTA L106V NP_001269363.1
NM_018645.5 569 Missense Mutation GCT,GGT A119G NP_061115.2
Gene
LOC101927958
Gene Name
uncharacterized LOC101927958
There are no transcripts associated with this gene.

Gene
LOC151174
Gene Name
uncharacterized LOC151174
There are no transcripts associated with this gene.

Gene
LOC643387
Gene Name
TAR DNA binding protein pseudogene
There are no transcripts associated with this gene.

Gene
PER2
Gene Name
period circadian clock 2
There are no transcripts associated with this gene.

View Full Product Details