Product Details

SNP ID
rs148876416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96848321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTCAGCTGGATAGCAGGTCCC[G/T]CAGGTCACTGTTGCAAGGCAGCAGG
Phenotype
MIM: 610736
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD23 PubMed Links

Gene Details

Gene
ANKRD23
Gene Name
ankyrin repeat domain 23
There are no transcripts associated with this gene.

Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016466.5 564 Silent Mutation NP_057550.3

View Full Product Details