Product Details

SNP ID
rs148975364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3499063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACCGGTTCTCCCACAAACAGC[C/T]GCATGGCCTTCGTGTAGTTCTGGAA
Phenotype
MIM: 613400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADI1 PubMed Links

Gene Details

Gene
ADI1
Gene Name
acireductone dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306077.1 3260 Missense Mutation CAG,CGG Q141R NP_001293006.1
NM_018269.3 3260 Missense Mutation CAG,CGG Q147R NP_060739.2

View Full Product Details