Product Details

SNP ID
rs149204343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241739754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGCGGCCAAGACTTATAGCCCA[A/G]TGAATAAAGTGAGAAGCCGAGAGCC
Phenotype
MIM: 609186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
D2HGDH PubMed Links

Gene Details

Gene
D2HGDH
Gene Name
D-2-hydroxyglutarate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287249.1 Intron NP_001274178.1
NM_152783.4 Intron NP_689996.4
XM_011511734.1 Intron XP_011510036.1
XM_011511735.1 Intron XP_011510037.1
XM_011511736.1 Intron XP_011510038.1
XM_011511737.2 Intron XP_011510039.1
XM_011511738.2 Intron XP_011510040.1
XM_011511739.1 Intron XP_011510041.1
XM_011511740.2 Intron XP_011510042.1
XM_011511741.2 Intron XP_011510043.1
XM_011511743.1 Intron XP_011510045.1
XM_011511744.1 Intron XP_011510046.1
XM_011511745.2 Intron XP_011510047.1
XM_011511746.2 Intron XP_011510048.1
XM_011511747.2 Intron XP_011510049.1
XM_011511749.2 Intron XP_011510051.1
XM_011511750.2 Intron XP_011510052.1
XM_011511751.1 Intron XP_011510053.1
XM_011511752.1 Intron XP_011510054.1
XM_011511753.2 Intron XP_011510055.1
XM_011511754.1 Intron XP_011510056.1
XM_011511756.1 Intron XP_011510058.1
XM_011511757.2 Intron XP_011510059.1
XM_011511758.2 Intron XP_011510060.1
XM_011511759.2 Intron XP_011510061.1
XM_011511760.2 Intron XP_011510062.1
XM_017004827.1 Intron XP_016860316.1
XM_017004828.1 Intron XP_016860317.1
XM_017004829.1 Intron XP_016860318.1
XM_017004830.1 Intron XP_016860319.1
XM_017004831.1 Intron XP_016860320.1
XM_017004832.1 Intron XP_016860321.1

View Full Product Details