Product Details

SNP ID
rs149250098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206767049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGTTTTTTGACTCAAAGCAGT[C/T]TCTTGTCCCTGTTGATAAATCTGAT
Phenotype
MIM: 612322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FASTKD2 PubMed Links

Gene Details

Gene
FASTKD2
Gene Name
FAST kinase domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136193.1 781 Missense Mutation TCT,TTT S119F NP_001129665.1
NM_001136194.1 781 Missense Mutation TCT,TTT S119F NP_001129666.1
NM_014929.3 781 Missense Mutation TCT,TTT S119F NP_055744.2
Gene
MDH1B
Gene Name
malate dehydrogenase 1B
There are no transcripts associated with this gene.

View Full Product Details