Product Details

SNP ID
rs149277361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24037839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTAGACAGCTGATAAGTTTC[C/G]TTTTCACTTTGTAAGTTCTTTCTTT
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 1787 Intron NP_001309892.1
NM_001322964.1 1787 Intron NP_001309893.1
NM_004116.4 1787 Intron NP_004107.1
NM_054033.3 1787 Intron NP_473374.1
XM_011532699.2 1787 Intron XP_011531001.1
XM_017003593.1 1787 Intron XP_016859082.1
XM_017003594.1 1787 Intron XP_016859083.1
XM_017003595.1 1787 Intron XP_016859084.1
XM_017003596.1 1787 Intron XP_016859085.1
Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 1787 Missense Mutation AAC,AAG N552K NP_001135791.1
NM_025203.2 1787 Missense Mutation AAC,AAG N552K NP_079479.1
XM_017005029.1 1787 Missense Mutation AAC,AAG N552K XP_016860518.1

View Full Product Details