Product Details

SNP ID
rs149353703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162319732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGACTGTGGGTGGTAGGCACATA[C/T]GCACCATAACTAGAATCTCATGGTC
Phenotype
MIM: 607030 MIM: 606951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCA PubMed Links

Gene Details

Gene
GCA
Gene Name
grancalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246446.2 Intron XP_005246503.1
XM_006712398.3 Intron XP_006712461.1
XM_006712400.3 Intron XP_006712463.1
XM_006712401.3 Intron XP_006712464.1
XM_011510926.2 Intron XP_011509228.1
XM_011510927.2 Intron XP_011509229.1
XM_011510928.2 Intron XP_011509230.1
XM_017003764.1 Intron XP_016859253.1
XM_017003765.1 Intron XP_016859254.1
XM_017003766.1 Intron XP_016859255.1
XM_017003767.1 Intron XP_016859256.1
XM_017003768.1 Intron XP_016859257.1
XM_017003769.1 Intron XP_016859258.1
Gene
IFIH1
Gene Name
interferon induced with helicase C domain 1
There are no transcripts associated with this gene.

View Full Product Details