Product Details

SNP ID
rs149369428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:107823417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCAAAAAGCCTATGGTCTATCCC[A/T]CATGACTGTCGGGAAGGCAATGTCA
Phenotype
MIM: 612707
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GACAT1 PubMed Links

Gene Details

Gene
GACAT1
Gene Name
gastric cancer associated transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RGPD4
Gene Name
RANBP2-like and GRIP domain containing 4
There are no transcripts associated with this gene.

Gene
RGPD4-AS1
Gene Name
RGPD4 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details