Product Details

SNP ID
rs149488308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176083412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGGGGGAGCGAAGCGCGGAG[C/G]AGCCCACCTCCACGTCGCTGCTCAT
Phenotype
MIM: 142991 MIM: 142989
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EVX2 PubMed Links

Gene Details

Gene
EVX2
Gene Name
even-skipped homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080458.1 551 Missense Mutation TCC,TGC S122C NP_001073927.1
Gene
HOXD13
Gene Name
homeobox D13
There are no transcripts associated with this gene.

View Full Product Details