Product Details

SNP ID
rs149785651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206745629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGTCTAAGAGCTTACCTGATTG[A/G]TATGGCTGAAAATGTATCTTGTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDH1B PubMed Links

Gene Details

Gene
MDH1B
Gene Name
malate dehydrogenase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039845.2 1665 Silent Mutation TAC,TAT Y467Y NP_001034934.1
NM_001282940.1 1665 Silent Mutation TAC,TAT Y467Y NP_001269869.1
XM_005246315.4 1665 Silent Mutation TAC,TAT Y369Y XP_005246372.1
XM_005246316.4 1665 Silent Mutation TAC,TAT Y254Y XP_005246373.1
XM_011510628.2 1665 Silent Mutation TAC,TAT Y309Y XP_011508930.1
XM_017003373.1 1665 Silent Mutation TAC,TAT Y369Y XP_016858862.1
XM_017003374.1 1665 Silent Mutation TAC,TAT Y309Y XP_016858863.1

View Full Product Details