Product Details
- SNP ID
-
rs150041202
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68646223 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGCTCTCCTGGGAGCACGGCCA[C/T]GTCCTGTGCACCTCTGTCAACTACC
- Phenotype
-
MIM: 607122
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROKR1
PubMed Links
Gene Details
- Gene
- PROKR1
- Gene Name
- prokineticin receptor 1
There are no transcripts associated with this gene.
View Full Product Details