Product Details

SNP ID
rs150099923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197462681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGAAATTGGATTTCCACCTGTG[C/T]TGGAGCGATATACATCAGTAGTAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COQ10B PubMed Links

Gene Details

Gene
COQ10B
Gene Name
coenzyme Q10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320818.1 579 Silent Mutation CTG,TTG L83L NP_001307747.1
NM_001320819.1 579 Silent Mutation CTG,TTG L105L NP_001307748.1
NM_001320820.1 579 Silent Mutation CTG,TTG L90L NP_001307749.1
NM_025147.4 579 Silent Mutation CTG,TTG L133L NP_079423.1

View Full Product Details