Product Details

SNP ID
rs150112597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176189453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAAGAGGAATGCCTCTAAGAAA[C/G]GTAAGTCCGCGGGCCTTGGATGGGG
Phenotype
MIM: 142987
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAGLR PubMed Links

Gene Details

Gene
HAGLR
Gene Name
HOXD antisense growth-associated long non-coding RNA
There are no transcripts associated with this gene.

Gene
HOXD1
Gene Name
homeobox D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024501.2 783 Missense Mutation CGC,GGC R218G NP_078777.1
XM_005246507.3 783 Missense Mutation CAC,GAC H218D XP_005246564.1
Gene
MIR7704
Gene Name
microRNA 7704
There are no transcripts associated with this gene.

View Full Product Details