Product Details

SNP ID
rs150209862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:180984026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATATCAAATCCTTTTTGTCTCT[G/T]CTCATTTCACTAACAGAATTCAGAA
Phenotype
MIM: 604151
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2E3 PubMed Links

Gene Details

Gene
UBE2E3
Gene Name
ubiquitin conjugating enzyme E2 E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278554.1 Intron NP_001265483.1
NM_001278555.1 Intron NP_001265484.1
NM_006357.3 Intron NP_006348.1
NM_182678.2 Intron NP_872619.1
XM_005246244.2 Intron XP_005246301.1
XM_011510492.2 Intron XP_011508794.1
XM_017003166.1 Intron XP_016858655.1
XM_017003167.1 Intron XP_016858656.1

View Full Product Details