Product Details

SNP ID
rs150242792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203706742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAACTTATTCCCTTCAATTCAA[A/G]TAACAGGTAAACAATGTTAATGTCT
Phenotype
MIM: 186760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD28 PubMed Links

Gene Details

Gene
CD28
Gene Name
CD28 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243077.1 268 Missense Mutation ATA,GTA I16V NP_001230006.1
NM_001243078.1 268 Missense Mutation ATA,GTA I16V NP_001230007.1
NM_006139.3 268 Missense Mutation ATA,GTA I16V NP_006130.1
XM_011512194.2 268 Intron XP_011510496.1
XM_011512195.2 268 Intron XP_011510497.1
XM_011512197.2 268 Missense Mutation ATA,GTA I16V XP_011510499.1

View Full Product Details