Product Details

SNP ID
rs150318966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208121822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGGACCCAGGAGCCCTCCAGCA[C/T]GTTGAGGGAGTGGATTTCATTGAAG
Phenotype
MIM: 123680 MIM: 123690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYGC PubMed Links

Gene Details

Gene
CRYGC
Gene Name
crystallin gamma C
There are no transcripts associated with this gene.

Gene
CRYGD
Gene Name
crystallin gamma D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006891.3 492 Missense Mutation ATG,GTG M126V NP_008822.2
Gene
LOC100507443
Gene Name
uncharacterized LOC100507443
There are no transcripts associated with this gene.

View Full Product Details