Product Details

SNP ID
rs150343979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25161217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCTGCCCCAGCGGAAGTGCTCC[A/G]TCCTGTAGGGGCCCTCGTCCTTCTT
Phenotype
MIM: 616797 MIM: 176830
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFR3B PubMed Links

Gene Details

Gene
EFR3B
Gene Name
EFR3 homolog B
There are no transcripts associated with this gene.

Gene
POMC
Gene Name
proopiomelanocortin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000939.3 1072 Missense Mutation ACG,ATG T223M NP_000930.1
NM_001035256.2 1072 Missense Mutation ACG,ATG T223M NP_001030333.1
NM_001319204.1 1072 Missense Mutation ACG,ATG T223M NP_001306133.1
NM_001319205.1 1072 Missense Mutation ACG,ATG T223M NP_001306134.1

View Full Product Details