Product Details

SNP ID
rs150386004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127863402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCCTCTCGAGCAGCACAGGAC[A/G]GTGTGGAGCAAACCCACTCACCCTT
Phenotype
MIM: 606017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMMECR1L PubMed Links

Gene Details

Gene
AMMECR1L
Gene Name
AMMECR1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199140.1 3484 UTR 3 NP_001186069.1
NM_031445.2 3484 Intron NP_113633.2
XM_005263804.4 3484 UTR 3 XP_005263861.1
XM_005263806.4 3484 UTR 3 XP_005263863.1
XM_005263807.4 3484 UTR 3 XP_005263864.1
XM_011511952.2 3484 UTR 3 XP_011510254.1
XM_011511954.2 3484 UTR 3 XP_011510256.1
XM_011511955.2 3484 Intron XP_011510257.1
XM_011511956.2 3484 Intron XP_011510258.1
XM_017005063.1 3484 Intron XP_016860552.1
Gene
LOC107985803
Gene Name
uncharacterized LOC107985803
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017005446.1 3484 Intron XP_016860935.1
Gene
POLR2D
Gene Name
RNA polymerase II subunit D
There are no transcripts associated with this gene.

View Full Product Details