Product Details

SNP ID
rs150480687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231108575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTCATTTTCAGTGAGGAGAAGC[A/G]TATCTAGTAGAATGATTGATGAAGA
Phenotype
MIM: 601122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR2B PubMed Links

Gene Details

Gene
HTR2B
Gene Name
5-hydroxytryptamine receptor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000867.4 1843 Missense Mutation NP_000858.3
NM_001320758.1 1843 Missense Mutation NP_001307687.1
XM_005246520.3 1843 Missense Mutation XP_005246577.1
XM_006712482.2 1843 Missense Mutation XP_006712545.1
Gene
PSMD1
Gene Name
proteasome 26S subunit, non-ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191037.1 1843 Intron NP_001177966.1
NM_002807.3 1843 Intron NP_002798.2
XM_017004517.1 1843 Intron XP_016860006.1

View Full Product Details