Product Details
- SNP ID
-
rs150488966
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:17543707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTAAAGCTTTTAAAGCATTTGC[A/G]TAGCTTTTCCTCAGGCAGATCTTTT
- Phenotype
-
MIM: 600817
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VSNL1
PubMed Links
Gene Details
- Gene
- VSNL1
- Gene Name
- visinin like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003385.4 |
|
Intron |
|
|
NP_003376.2 |
View Full Product Details