Product Details

SNP ID
rs150519950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85549976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATCAGGATTTGACTCAGGAGG[A/G]TTAGAATGTGAAGAATCCGTGTTTG
Phenotype
MIM: 137167 MIM: 601468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGCX PubMed Links

Gene Details

Gene
GGCX
Gene Name
gamma-glutamyl carboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000821.6 2157 Silent Mutation AAC,AAT N745N NP_000812.2
NM_001142269.3 2157 Silent Mutation AAC,AAT N688N NP_001135741.1
NM_001311312.1 2157 Intron NP_001298241.1
XM_005264259.4 2157 Silent Mutation AAC,AAT N743N XP_005264316.1
XM_011532764.2 2157 Silent Mutation AAC,AAT N471N XP_011531066.1
XM_011532765.2 2157 Silent Mutation AAC,AAT N471N XP_011531067.1
XM_017003803.1 2157 Silent Mutation AAC,AAT N686N XP_016859292.1
Gene
MAT2A
Gene Name
methionine adenosyltransferase 2A
There are no transcripts associated with this gene.

View Full Product Details