Product Details

SNP ID
rs150780439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:40080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATAGTACTGAGTACTGAATTG[A/C]GTGGCAAACACTCCACTTGCAGTCA
Phenotype
MIM: 611395
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM110C PubMed Links

Gene Details

Gene
FAM110C
Gene Name
family with sequence similarity 110 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077710.2 2755 UTR 3 NP_001071178.2
XM_011510372.2 2755 UTR 3 XP_011508674.1
XM_017004689.1 2755 Intron XP_016860178.1
XM_017004690.1 2755 Intron XP_016860179.1
XM_017004691.1 2755 UTR 3 XP_016860180.1
XM_017004692.1 2755 Intron XP_016860181.1

View Full Product Details