Product Details

SNP ID
rs150861176
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112723260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCAGGCCTCCAGTCCCCATAG[C/T]GTTTCCATCCTCTTCCTACTGAGGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NT5DC4 PubMed Links

Gene Details

Gene
NT5DC4
Gene Name
5'-nucleotidase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011512262.2 Intron XP_011510564.1
XM_011512265.2 Intron XP_011510567.1
XM_017005474.1 Intron XP_016860963.1
XM_017005475.1 Intron XP_016860964.1
XM_017005476.1 Intron XP_016860965.1
XM_017005477.1 Intron XP_016860966.1
XM_017005478.1 Intron XP_016860967.1

View Full Product Details