Product Details

SNP ID
rs150938897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74221491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGAGTGAGTAACTCCAACTGGA[A/G]GATCTTTTTCCTGGCAGGAAAATGA
Phenotype
MIM: 604887 MIM: 606757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTHFD2 PubMed Links

Gene Details

Gene
MTHFD2
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
There are no transcripts associated with this gene.

Gene
SLC4A5
Gene Name
solute carrier family 4 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021196.3 3740 Silent Mutation TCC,TCT S1130S NP_067019.3
NM_133478.2 3740 Silent Mutation TCC,TCT S1114S NP_597812.1

View Full Product Details