Product Details

SNP ID
rs151024127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240590956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCAGGGAGACTCTGTTTCTCC[C/T]GCTGCCAGAGGGAGGATGTGTTCTG
Phenotype
MIM: 605286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN10 PubMed Links

Gene Details

Gene
CAPN10
Gene Name
calpain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023083.3 611 Missense Mutation CGC,TGC R139C NP_075571.1
NM_023085.3 611 Missense Mutation CGC,TGC R139C NP_075573.2
Gene
CAPN10-AS1
Gene Name
CAPN10 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details