Product Details

SNP ID
rs151073603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70809274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCTGTGCTCAGGTTGCTCTTA[C/G]TTACTGAGGATCCAGGGACAGGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4F PubMed Links

Gene Details

Gene
CLEC4F
Gene Name
C-type lectin domain family 4 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258027.1 1844 Intron NP_001244956.1
NM_001321308.1 1844 Intron NP_001308237.1
NM_173535.2 1844 Missense Mutation AAC,AAG N589K NP_775806.2
XM_011532635.2 1844 Missense Mutation AAC,AAG N699K XP_011530937.1
XM_011532637.2 1844 Missense Mutation AAC,AAG N699K XP_011530939.1
XM_011532638.2 1844 Intron XP_011530940.1
XM_011532639.2 1844 Intron XP_011530941.1
XM_011532640.2 1844 Intron XP_011530942.1
XM_011532641.2 1844 Intron XP_011530943.1
XM_011532642.2 1844 Missense Mutation AAC,AAG N559K XP_011530944.1
XM_011532643.1 1844 Missense Mutation AAC,AAG N528K XP_011530945.1
XM_017003519.1 1844 Missense Mutation AAC,AAG N699K XP_016859008.1

View Full Product Details