Product Details

SNP ID
rs151106160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190883132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGTCAAACAGGAAATTGAGCT[G/T]CTTCTGTGATTCACTTGTCCAGAAG
Phenotype
MIM: 138280
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLS PubMed Links

Gene Details

Gene
GLS
Gene Name
glutaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256310.1 Intron NP_001243239.1
NM_014905.4 Intron NP_055720.3
XM_005246467.3 Intron XP_005246524.1
XM_006712435.1 Intron XP_006712498.1
XM_017003829.1 Intron XP_016859318.1

View Full Product Details