Product Details

SNP ID
rs151119195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73291504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCGAGTAAAAGCCGAGGCCCT[A/T]GAGCCGCTCCTCGGCGCGCGCCTTC
Phenotype
MIM: 128992 MIM: 609108
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EGR4 PubMed Links

Gene Details

Gene
EGR4
Gene Name
early growth response 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001965.3 1798 Nonsense Mutation AAG,TAG K575* NP_001956.3
Gene
FBXO41
Gene Name
F-box protein 41
There are no transcripts associated with this gene.

View Full Product Details