Product Details

SNP ID
rs151139937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27293535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAGACAAACAGGTCATTCTG[A/G]TTGAAAGCAGGGCCCTTATAGAGCT
Phenotype
MIM: 606474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM54 PubMed Links

Gene Details

Gene
TRIM54
Gene Name
tripartite motif containing 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032546.3 Intron NP_115935.3
NM_187841.2 Intron NP_912730.2

View Full Product Details