Product Details

SNP ID
rs151306502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88526495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATACTCCAGGGTCACGCCGCAA[C/G]AGGTAGGAAGGCACGCAAGTCCTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TEX37 PubMed Links

Gene Details

Gene
TEX37
Gene Name
testis expressed 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152670.2 235 Missense Mutation CAG,GAG Q40E NP_689883.1
XM_005264188.3 235 Missense Mutation CAG,GAG Q40E XP_005264245.3
XM_011532691.1 235 Missense Mutation CAG,GAG Q40E XP_011530993.1
XM_011532692.1 235 Missense Mutation CAG,GAG Q40E XP_011530994.1

View Full Product Details