Product Details

SNP ID
rs121918136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49022146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCGCCGCGCGTTGGTCAGCCA[C/T]GGCCACCGCTCTCGCGCTACGTAGC
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 606031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009996.2 506 Intron NP_001009996.1
NM_001276405.1 506 Intron NP_001263334.1
NM_018114.5 506 Intron NP_060584.3
XM_005265269.3 506 Intron XP_005265326.1
XM_006713219.2 506 Intron XP_006713282.1
XM_011533891.1 506 Intron XP_011532193.1
XM_011533892.1 506 Intron XP_011532194.1
XM_011533893.1 506 Intron XP_011532195.1
XM_017006723.1 506 Intron XP_016862212.1
Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199069.1 506 Missense Mutation ACG,ATG T1M NP_951032.1
NM_199070.1 506 Intron NP_951033.1
NM_199073.1 506 Intron NP_951047.1
NM_199074.1 506 Intron NP_951056.1
Gene
QRICH1
Gene Name
glutamine rich 1
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details