Product Details

SNP ID
rs121918652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:181712749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATAAGTACACGCTGCCCGGCG[C/G]GCTGCTGGCCCCCGGCGGCAATAGC
Phenotype
MIM: 184429 MIM: 616338
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SOX2 PubMed Links

Gene Details

Gene
SOX2
Gene Name
SRY-box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003106.3 826 Missense Mutation GCG,GGG A130G NP_003097.1
Gene
SOX2-OT
Gene Name
SOX2 overlapping transcript
There are no transcripts associated with this gene.

View Full Product Details