Product Details

SNP ID
rs137853065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38138978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGGCCGACTGCTCGAGCTGC[C/T]TACCAAGCTGGGCCGCGACGACGTG
Phenotype
MIM: 604054 MIM: 602170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
There are no transcripts associated with this gene.

Gene
MYD88
Gene Name
myeloid differentiation primary response 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172566.1 501 Missense Mutation CCT,CTT P106L NP_001166037.1
NM_001172567.1 501 Missense Mutation CCT,CTT P106L NP_001166038.1
NM_001172568.1 501 Missense Mutation CCT,CTT P106L NP_001166039.1
NM_001172569.1 501 Missense Mutation CCT,CTT P106L NP_001166040.1
NM_002468.4 501 Missense Mutation CCT,CTT P106L NP_002459.2
XM_005265172.1 501 Missense Mutation CCT,CTT P106L XP_005265229.1
XM_006713170.1 501 Missense Mutation CCT,CTT P106L XP_006713233.1

View Full Product Details