Product Details

SNP ID
rs137867732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184032960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCACAGGGTGGACCCTGCAGCC[G/T]TTCAAGCAGTGTTTGACAGAAAGGC
Phenotype
MIM: 610122
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR3D PubMed Links

Gene Details

Gene
HTR3D
Gene Name
5-hydroxytryptamine receptor 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145143.1 130 Intron NP_001138615.1
NM_001163646.1 130 Missense Mutation GTT,TTT V44F NP_001157118.1
NM_182537.2 130 Intron NP_872343.2
XM_017005854.1 130 Intron XP_016861343.1

View Full Product Details