Product Details

SNP ID
rs137924029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119586386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGCGGCTGTGGGGCTGCCATG[C/T]GGGCCATGTGCATCGGTCTCAGGTT
Phenotype
MIM: 603081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADPRH PubMed Links

Gene Details

Gene
ADPRH
Gene Name
ADP-ribosylarginine hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001125.3 521 Missense Mutation CGG,TGG R134W NP_001116.1
NM_001291949.1 521 Missense Mutation CGG,TGG R134W NP_001278878.1
NM_001291950.1 521 Missense Mutation CGG,TGG R27W NP_001278879.1
XM_011512438.2 521 Missense Mutation CGG,TGG R134W XP_011510740.1

View Full Product Details