Product Details

SNP ID
rs137965425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37994459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTCTGCAGCTACTTCCGCCCG[A/G]GAATCATGTGAGTGCGGGGGCGACC
Phenotype
MIM: 608592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 684 Missense Mutation AGA,GGA R112G NP_056957.3
XM_005265191.3 684 Missense Mutation AGA,GGA R112G XP_005265248.1
XM_005265192.4 684 Missense Mutation AGA,GGA R112G XP_005265249.1
XM_006713184.3 684 Missense Mutation AGA,GGA R112G XP_006713247.1
XM_011533770.2 684 Missense Mutation AGA,GGA R112G XP_011532072.1
XM_011533771.1 684 Missense Mutation AGA,GGA R112G XP_011532073.1
XM_011533773.2 684 Missense Mutation AGA,GGA R112G XP_011532075.1

View Full Product Details