Product Details

SNP ID
rs137975956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100364453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTTTAGCCACATGTCCTTCAC[A/G]AGAAATTTTAAGCTATTTTAATACT
Phenotype
MIM: 616769 MIM: 606081
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NIT2 PubMed Links

Gene Details

Gene
NIT2
Gene Name
nitrilase family member 2
There are no transcripts associated with this gene.

Gene
TOMM70
Gene Name
translocase of outer mitochondrial membrane 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014820.4 3387 UTR 3 NP_055635.3

View Full Product Details