Product Details
- SNP ID
-
rs137995657
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44864290 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTAGCCGGGGCCTCAGTTTCTCC[A/G]TGTCTTCAGCCATTGCATCTGTCAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR564
PubMed Links
Gene Details
- Gene
- MIR564
- Gene Name
- microRNA 564
There are no transcripts associated with this gene.
- Gene
- TMEM42
- Gene Name
- transmembrane protein 42
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144638.2 |
302 |
Missense Mutation |
ATG,GTG |
M96V |
NP_653239.1 |
View Full Product Details