Product Details

SNP ID
rs138004710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:147410440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCGCAACCGGGGTTTTGGCGAC[C/G]CGGCGGCGGCAGCCAGCGCACAGCA
Phenotype
MIM: 600470 MIM: 608948
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZIC1 PubMed Links

Gene Details

Gene
ZIC1
Gene Name
Zic family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003412.3 1047 Missense Mutation CCG,GCG P110A NP_003403.2
Gene
ZIC4
Gene Name
Zic family member 4
There are no transcripts associated with this gene.

View Full Product Details