Product Details

SNP ID
rs138181654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101651388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTACCACAGTCTCAAGTTCTTG[A/G]GGACCACTGCTTTCTTGTTCCTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB11 PubMed Links

Gene Details

Gene
ZBTB11
Gene Name
zinc finger and BTB domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014415.3 4555 Silent Mutation CCC,CCT P980P NP_055230.2
XM_011512689.2 4555 Silent Mutation CCC,CCT P915P XP_011510991.1
XM_011512690.2 4555 Silent Mutation CCC,CCT P696P XP_011510992.1

View Full Product Details