Product Details

SNP ID
rs138217463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193277778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTGCCACCACTCCAATCCAGA[A/G]CTAACTCTTCTCCATCCCTTGACCT
Phenotype
MIM: 606487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP13A5 PubMed Links

Gene Details

Gene
ATP13A5
Gene Name
ATPase 13A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198505.2 Intron NP_940907.2
XM_011512770.1 Intron XP_011511072.1
XM_017006304.1 Intron XP_016861793.1
XM_017006305.1 Intron XP_016861794.1
Gene
HRASLS
Gene Name
HRAS like suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020386.4 Intron NP_065119.2
XM_011513034.1 Intron XP_011511336.1
XM_011513035.2 Intron XP_011511337.1
XM_011513037.2 Intron XP_011511339.1
XM_017006923.1 Intron XP_016862412.1

View Full Product Details