Product Details

SNP ID
rs138294030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122414833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTATAATCCCATAGTTTAAGAGT[A/G]TTGTCCAAAGTTGCAGTGAGAATGT
Phenotype
MIM: 608017 MIM: 600686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM162A PubMed Links

Gene Details

Gene
FAM162A
Gene Name
family with sequence similarity 162 member A
There are no transcripts associated with this gene.

Gene
KPNA1
Gene Name
karyopherin subunit alpha 1
There are no transcripts associated with this gene.

Gene
LOC102723582
Gene Name
uncharacterized LOC102723582
There are no transcripts associated with this gene.

Gene
WDR5B
Gene Name
WD repeat domain 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019069.3 1203 Silent Mutation AAC,AAT N232N NP_061942.2

View Full Product Details