Product Details

SNP ID
rs138369951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:147412609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCACATGCACGTGCACACGAG[C/T]GACAAGCCCTATCTTTGCAAGATGT
Phenotype
MIM: 600470 MIM: 608948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC440982 PubMed Links

Gene Details

Gene
LOC440982
Gene Name
uncharacterized LOC440982
There are no transcripts associated with this gene.

Gene
ZIC1
Gene Name
Zic family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003412.3 1793 Silent Mutation AGC,AGT S358S NP_003403.2
Gene
ZIC4
Gene Name
Zic family member 4
There are no transcripts associated with this gene.

View Full Product Details