Product Details

SNP ID
rs138659151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:71771925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGAGACTAAGGAAAAAAAACTT[C/T]ATAAAAAGTAAAAGAAGTAAATGTA
Phenotype
MIM: 607002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROK2 PubMed Links

Gene Details

Gene
PROK2
Gene Name
prokineticin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126128.1 1280 UTR 3 NP_001119600.1
NM_021935.3 1280 UTR 3 NP_068754.1
XM_017006974.1 1280 UTR 3 XP_016862463.1

View Full Product Details