Product Details

SNP ID
rs138785691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179592676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTCCACATAAGGCAAGGCAAA[A/G]AATCGTTTCCTATTGTATCTTTTAT
Phenotype
MIM: 604958 MIM: 611852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL6A PubMed Links

Gene Details

Gene
ACTL6A
Gene Name
actin like 6A
There are no transcripts associated with this gene.

Gene
MRPL47
Gene Name
mitochondrial ribosomal protein L47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020409.2 473 Silent Mutation TTC,TTT F199F NP_065142.2
NM_177988.1 473 Silent Mutation TTC,TTT F89F NP_817125.1

View Full Product Details