Product Details
- SNP ID
-
rs138846385
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:63823487 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAAGAAGCCCTCCTGTCAAACAC[A/G]CAGAGCCTTCTCCCTAGGATTGTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf49
PubMed Links
Gene Details
- Gene
- C3orf49
- Gene Name
- chromosome 3 open reading frame 49
There are no transcripts associated with this gene.
View Full Product Details