Product Details

SNP ID
rs138996733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196324371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGGACCATCTTTCAGAGCAA[C/T]TCCAGAAGTGACAAAGCAAATCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCTEX1D2 PubMed Links

Gene Details

Gene
TCTEX1D2
Gene Name
Tctex1 domain containing 2
There are no transcripts associated with this gene.

Gene
TM4SF19
Gene Name
transmembrane 4 L six family member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204897.1 507 Missense Mutation ATT,GTT I117V NP_001191826.1
NM_001204898.1 507 Missense Mutation ATT,GTT I91V NP_001191827.1
NM_138461.3 507 Missense Mutation ATT,GTT I117V NP_612470.2
Gene
TM4SF19-AS1
Gene Name
TM4SF19 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TM4SF19-TCTEX1D2
Gene Name
TM4SF19-TCTEX1D2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details