Product Details

SNP ID
rs139023558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169793769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTGCAAGATATACACGTCCAT[C/T]TACCACAAATGGCTCCACATCTGTA
Phenotype
MIM: 606042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172779.1 1229 Missense Mutation AAT,GAT N421D NP_001166250.1
NM_001172780.1 1229 Intron NP_001166251.1
NM_153353.4 1229 Missense Mutation AAT,GAT N389D NP_699184.2
XM_006713508.3 1229 Missense Mutation AAT,GAT N403D XP_006713571.1
XM_011512442.2 1229 Missense Mutation AAT,GAT N420D XP_011510744.1
XM_017005746.1 1229 Missense Mutation AAT,GAT N359D XP_016861235.1
Gene
MYNN
Gene Name
myoneurin
There are no transcripts associated with this gene.

View Full Product Details