Product Details

SNP ID
rs139425191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49016167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGGGACACTCACCTCATCATG[C/T]AGCAGTGAGAAGTCCAGACTGCTCA
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 614584 MIM: 606031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009996.2 1696 Silent Mutation CTA,CTG L440L NP_001009996.1
NM_001276405.1 1696 Silent Mutation CTA,CTG L440L NP_001263334.1
NM_018114.5 1696 Silent Mutation CTA,CTG L273L NP_060584.3
XM_005265269.3 1696 Silent Mutation CTA,CTG L440L XP_005265326.1
XM_006713219.2 1696 Silent Mutation CTA,CTG L273L XP_006713282.1
XM_011533891.1 1696 Silent Mutation CTA,CTG L440L XP_011532193.1
XM_011533892.1 1696 Silent Mutation CTA,CTG L273L XP_011532194.1
XM_011533893.1 1696 UTR 3 XP_011532195.1
XM_017006723.1 1696 Silent Mutation CTA,CTG L273L XP_016862212.1
Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320546.1 1696 Intron NP_001307475.1
NM_001320547.1 1696 Intron NP_001307476.1
NM_018031.4 1696 Intron NP_060501.3

View Full Product Details