Product Details

SNP ID
rs139657148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9816024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGGGAGAGGCTGCCTTAGGC[A/C]CTGCTACCCACACTGGCCTCTGTGT
Phenotype
MIM: 604226
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARPC4 PubMed Links

Gene Details

Gene
ARPC4
Gene Name
actin related protein 2/3 complex subunit 4
There are no transcripts associated with this gene.

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198793.1 Intron NP_001185722.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025930.3 Intron NP_001021100.3

View Full Product Details